siRNA Details
Gene Symbol: Rnf19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013923.2 | NP_038951.1 | 10 | 3703 | ||
GenBank mRNA | AF120207.1 | - | 9 | 3596 | ||
AK082665.1 | - | 8 | 3297 | |||
AK147768.1 | - | 10 | 3691 | |||
BC040769.1 | - | 9 | 3478 | 4457807 | ||
X71642.1 | - | 6 | 2248 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001130560, FQ230338.
Panther Classification:
Gene Ontology Categories:
