siRNA Details
| Gene Symbol: Neu2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160163.1 | NP_001153635.1 | 3 | 390 | ||
| NM_001160164.1 | NP_001153636.1 | 2 | 288 | |||
| NM_001160165.1 | NP_001153637.1 | 2 | 416 | |||
| NM_015750.3 | NP_056565.1 | 2 | 256 | |||
| GenBank mRNA | AB028023.1 | - | 2 | 258 | ||
| AB048604.1 | - | 2 | 513 | |||
| AF139059.1 | - | 2 | 370 | |||
| AK009828.1 | - | 1 | 470 | |||
| AK145284.1 | - | 2 | 289 | |||
| BC026408.1 | - | 1 | 44 | |||
| BC105657.1 | - | 2 | 258 | 40046586 | ||
| BC105658.1 | - | 2 | 258 | 40046587 | ||
| BC105659.1 | - | 2 | 258 | 40046588 | ||
| BC105661.1 | - | 1 | 518 | 40046584 | ||
| FJ613504.1 | - | 3 | 373 | |||

Neu2
Genomic Map