siRNA Details
| Gene Symbol: Shroom3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015756.2 | NP_056571.2 | 2 | 507 | ||
| GenBank mRNA | AF199421.1 | - | 2 | 507 | ||
| BC145113.1 | - | 2 | 317 | 9053661 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shroom3| Gene Symbol: Shroom3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015756.2 | NP_056571.2 | 2 | 507 | ||
| GenBank mRNA | AF199421.1 | - | 2 | 507 | ||
| BC145113.1 | - | 2 | 317 | 9053661 | ||