siRNA Details
| Gene Symbol: Postn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198765.1 | NP_001185694.1 | 4 | 432 | ||
| NM_001198766.1 | NP_001185695.1 | 4 | 432 | |||
| NM_015784.3 | NP_056599.1 | 4 | 432 | |||
| GenBank mRNA | AK030756.1 | - | 4 | 431 | ||
| AK154049.1 | - | 4 | 430 | |||
| AY651928.1 | - | 4 | 402 | |||
| BC007141.1 | - | 4 | 425 | |||
| D13664.1 | - | 4 | 414 | |||
| E05655.1 | - | 4 | 428 | |||
| KM117171.1 | - | 4 | 396 | |||
| KM117172.1 | - | 4 | 396 | |||

Postn
Genomic Map