siRNA Details
| Gene Symbol: Crym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016669.1 | NP_057878.1 | 2 | 348 | ||
| GenBank mRNA | AF039391.1 | - | 2 | 348 | ||
| AK138422.1 | - | 2 | 319 | |||
| AK143116.1 | - | 2 | 338 | |||
| AK163781.1 | - | 2 | 319 | |||
| AK163801.1 | - | 2 | 345 | |||
| BC045159.1 | - | 2 | 309 | 6492374 | ||

Crym
Genomic Map