siRNA Details
| Gene Symbol: Hpcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016677.4 | NP_057886.1 | 2 | 454 | ||
| GenBank mRNA | AF085192.1 | - | 2 | 457 | ||
| AK002849.1 | - | 2 | 437 | |||
| BC001997.1 | - | 2 | 430 | 3496729 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hpcal1| Gene Symbol: Hpcal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016677.4 | NP_057886.1 | 2 | 454 | ||
| GenBank mRNA | AF085192.1 | - | 2 | 457 | ||
| AK002849.1 | - | 2 | 437 | |||
| BC001997.1 | - | 2 | 430 | 3496729 | ||