siRNA Details
| Gene Symbol: Zeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289521.1 | NP_001276450.1 | 2 | 320 | ||
| NM_015753.4 | NP_056568.2 | 3 | 657 | |||
| GenBank mRNA | AF033116.1 | - | 2 | 172 | ||
| AK012377.1 | - | 2 | 128 | |||
| AK031541.1 | - | 4 | 754 | |||
| AK122312.1 | - | 3 | 230 | |||
| AK135557.1 | - | 3 | 467 | |||
| AK141157.1 | - | 2 | 288 | |||
| AK157561.1 | - | 3 | 380 | |||
| BC060699.1 | - | 3 | 394 | 6408845 | ||
| CB245498.1 | - | 3 | 657 | |||
| CJ130482.1 | - | 2 | 320 | |||

Zeb2
Genomic Map