siRNA Details
| Gene Symbol: Greb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252071.1 | NP_001239000.1 | 4 | 613 | ||
| NM_015764.4 | NP_056579.2 | 6 | 935 | |||
| GenBank mRNA | AF180470.1 | - | 5 | 591 | ||
| AK045057.1 | - | 3 | 389 | |||
| AK147337.1 | - | 4 | 463 | |||
| AK157808.1 | - | 4 | 547 | |||
| BC160931.1 | - | 5 | 545 | |||

Greb1
Genomic Map