siRNA Details
| Gene Symbol: Ppap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005452.1 | - | 3 | 486 | ||
| AK049581.1 | - | 3 | 529 | |||
| AK179023.1 | - | 2 | 290 | |||
| AK208995.1 | - | 2 | 235 | |||
| BC010332.1 | - | 3 | 507 | 2654393 | ||
| BC085104.1 | - | 3 | 445 | |||
| BQ571983.1 | - | 2 | 332 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF123611.1 | - | 521 | Chr. - - [-] on Build | ||

Ppap2c
Genomic Map