siRNA Details
| Gene Symbol: Fbxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015822.2 | NP_056637.1 | 2 | 562 | ||
| GenBank mRNA | AF176521.1 | - | 2 | 269 | ||
| AK080863.1 | - | 2 | 571 | |||
| AK142007.1 | - | 2 | 562 | |||
| AK166128.1 | - | 2 | 443 | |||
| AK167409.1 | - | 2 | 381 | |||
| BC026835.1 | - | 1 | 228 | |||
| BC037665.1 | - | 1 | 225 | |||
| BC067203.1 | - | 2 | 444 | 6334317 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001100568, XM_006252442, CB718262.

Fbxl3
Genomic Map