siRNA Details
| Gene Symbol: Postn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198765.1 | NP_001185694.1 | 22 | 3080 | ||
| NM_001198766.1 | NP_001185695.1 | 21 | 2999 | |||
| NM_015784.3 | NP_056599.1 | 22 | 3083 | |||
| GenBank mRNA | AK154049.1 | - | 21 | 2997 | ||
| BC007141.1 | - | 22 | 3074 | |||
| BC031449.1 | - | 21 | 2968 | 4457222 | ||
| BE850247.1 | - | 1 | 215 | |||
| D13664.1 | - | 22 | 3065 | |||
| E05655.1 | - | 22 | 3079 | |||

Postn
Genomic Map