siRNA Details
Gene Symbol: Fbxo18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015792.1 | NP_056607.1 | 19 | 2836 | ||
GenBank mRNA | AF184275.1 | - | 17 | 2330 | ||
AK003964.1 | - | 4 | 397 | |||
AK084645.1 | - | 19 | 2780 | |||
AK147829.1 | - | 19 | 2820 | |||
AK154357.1 | - | 19 | 2859 | |||
AK158937.1 | - | 19 | 2887 | |||
AK215165.1 | - | 153 | ||||
BC004036.1 | - | 16 | 1922 | |||
BC031393.1 | - | 19 | 2836 | 4948804 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
