siRNA Details
| Gene Symbol: Dkk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015814.2 | NP_056629.1 | 8 | 2169 | ||
| GenBank mRNA | AF177400.1 | - | 8 | 2156 | ||
| AJ243964.1 | - | 8 | 2172 | |||
| AK004853.1 | - | 8 | 2169 | |||
| AK013054.2 | - | 7 | 2110 | |||
| AK013622.1 | - | 7 | 2081 | |||
| BC005519.1 | - | 7 | 1869 | |||
| BC046304.1 | - | 7 | 2084 | 6493718 | ||
| BC050934.1 | - | 7 | 2066 | 6812621 | ||

Dkk3
Genomic Map