siRNA Details
| Gene Symbol: Klra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016659.3 | NP_057868.2 | 9 | 1493 | ||
| GenBank mRNA | AK080158.1 | - | 7 | 1075 | ||
| AK172530.1 | - | 976 | ||||
| BC140968.1 | - | 6 | 969 | 9055580 | ||
| BC144929.1 | - | 5 | 879 | 9053477 | ||
| M25775.1 | - | 7 | 1097 | |||
| M25812.1 | - | 7 | 1031 | |||

Klra1
Genomic Map