siRNA Details
| Gene Symbol: Sfrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016687.3 | NP_057896.1 | 1 | 686 | ||
| GenBank mRNA | AF117709.1 | - | 1 | 701 | ||
| AK009981.1 | - | 1 | 684 | |||
| AK080766.1 | - | 1 | 687 | |||
| BC034853.1 | - | 1 | 645 | 1364770 | ||
More Information
Set Membership: |
|

Sfrp4
Genomic Map