siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016704.2 | NP_057913.2 | 3, 4 | 436 | ||
| GenBank mRNA | AF184900.1 | - | 3, 4 | 437 | ||
| AK041057.1 | - | 3, 4 | 446 | |||
| BB222000.2 | - | 2, 3 | 188 | |||
| BC011251.1 | - | 3, 4 | 425 | 4196330 | ||

C6
Genomic Map