siRNA Details
| Gene Symbol: Crlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164735.1 | NP_001158207.1 | 8 | 1054 | ||
| NM_016715.4 | NP_057924.3 | 8 | 1021 | |||
| GenBank mRNA | AB031333.1 | - | 8 | 969 | ||
| AB039945.1 | - | 8 | 917 | |||
| AF201963.1 | - | 8 | 1105 | |||
| AF232936.1 | - | 8 | 985 | |||
| AK010291.1 | - | 3 | 213 | |||
| AK149928.1 | - | 8 | 1023 | |||
| AK170466.1 | - | 5 | 883 | |||
| BC016580.1 | - | 8 | 1099 | 2655857 | ||
| BC023788.1 | - | 8 | 1009 | 5346748 | ||

Crlf2
Genomic Map