siRNA Details
| Gene Symbol: Cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016756.4 | NP_058036.1 | 2 | 359 | ||
| NM_183417.3 | NP_904326.1 | 2 | 359 | |||
| GenBank mRNA | AJ223732.1 | - | 2 | 261 | ||
| AK036589.1 | - | 1 | 1020 | |||
| AK042462.1 | - | 1 | 1020 | |||
| AK146845.1 | - | 2 | 339 | |||
| AK148017.1 | - | 2 | 298 | |||
| AK151979.1 | - | 2 | 309 | |||
| AK152922.1 | - | 2 | 312 | |||
| AK153983.1 | - | 2 | 311 | |||
| AK154997.1 | - | 2 | 311 | |||
| AK179714.1 | - | 1 | 30 | |||
| AK181235.1 | - | 1 | 30 | |||
| AK186022.1 | - | 1 | 30 | |||
| AK186779.1 | - | 1 | 30 | |||
| AK187629.1 | - | 2 | 95 | |||
| AK189612.1 | - | 2 | 95 | |||
| AK210959.1 | - | 1 | 30 | |||
| AK219393.1 | - | 1 | 30 | |||
| BC005654.1 | - | 2 | 320 | 3709730 | ||
| BC026145.1 | - | 1 | 1065 | |||
| CN705407.1 | - | 2 | 359 | |||
| U63337.1 | - | 2 | 287 | |||

Cdk2
Genomic Map