siRNA Details
| Gene Symbol: KCNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004770.2 | NP_004761.2 | 2 | 700 | ||
| GenBank mRNA | AF338730.1 | - | 2 | 700 | ||
| AF450111.1 | - | 1 | 112 | |||
| U69962.1 | - | 1 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNB2| Gene Symbol: KCNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004770.2 | NP_004761.2 | 2 | 700 | ||
| GenBank mRNA | AF338730.1 | - | 2 | 700 | ||
| AF450111.1 | - | 1 | 112 | |||
| U69962.1 | - | 1 | 114 | |||