siRNA Details
| Gene Symbol: Sult1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016771.3 | NP_058051.3 | 3 | 442 | ||
| GenBank mRNA | AF026073.1 | - | 3 | 320 | ||
| AI118913.1 | - | 3 | 454 | |||
| AK133530.1 | - | 3 | 361 | |||
| AK160466.1 | - | 3 | 427 | |||
| BC010752.1 | - | 3 | 415 | |||
| BC066190.1 | - | 3 | 395 | 30472198 | ||
| U32371.1 | - | 3 | 392 | |||

Sult1d1
Genomic Map