siRNA Details
| Gene Symbol: Pgrmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016783.3 | NP_058063.2 | 1 | 358 | ||
| GenBank mRNA | AF042491.1 | - | 1 | 309 | ||
| AK153272.1 | - | 1 | 375 | |||
| AK159101.1 | - | 1 | 375 | |||
| AK159345.1 | - | 1 | 375 | |||
| AK159599.1 | - | 1 | 375 | |||
| AK162315.1 | - | 1 | 378 | |||
| AK166240.1 | - | 1 | 375 | |||
| AK167655.1 | - | 1 | 375 | |||
| AK168387.1 | - | 1 | 374 | |||
| AK168882.1 | - | 1 | 375 | |||
| AK169063.1 | - | 2 | 318 | |||
| BC006016.1 | - | 1 | 358 | 3591536 | ||

Pgrmc1
Genomic Map