siRNA Details
| Gene Symbol: Neu3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016720.2 | NP_057929.1 | 3 | 1108 | ||
| GenBank mRNA | AB026842.1 | - | 3 | 1107 | ||
| AK050428.1 | - | 3 | 986 | |||
| AK134405.1 | - | 3 | 1065 | |||
| BC116892.1 | - | 3 | 897 | 40126211 | ||
| BC116924.1 | - | 3 | 897 | 40126243 | ||

Neu3
Genomic Map