siRNA Details
| Gene Symbol: Galns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193645.1 | NP_001180574.1 | 3, 4 | 462 | ||
| NM_016722.4 | NP_057931.3 | 3, 4 | 462 | |||
| GenBank mRNA | AF111346.1 | - | 3, 4 | 383 | ||
| AK033946.1 | - | 3, 4 | 380 | |||
| AK159592.1 | - | 3, 4 | 439 | |||
| AK220245.1 | - | 3, 4 | 364 | |||
| BC004002.1 | - | 3, 4 | 316 | 3492664 | ||
| BQ550288.1 | - | 3, 4 | 228 | |||

Galns
Genomic Map