siRNA Details
| Gene Symbol: KCNJ8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004982.3 | NP_004973.1 | 2, 3 | 728 | ||
| XM_005253358.4 | XP_005253415.1 | 3, 4 | 697 | |||
| XM_017019283.1 | XP_016874772.1 | 3, 4 | 632 | |||
| XM_017019284.1 | XP_016874773.1 | 3, 4 | 703 | |||
| GenBank mRNA | AK291821.1 | - | 3, 4 | 536 | ||
| AL546327.3 | - | 2, 3 | 728 | |||
| BC000544.2 | - | 2, 3 | 588 | 3161776 | ||
| D50312.1 | - | 2, 3 | 652 | |||

KCNJ8
Genomic Map