siRNA Details
| Gene Symbol: Smad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016769.4 | NP_058049.3 | 9 | 1718 | ||
| GenBank mRNA | AF016189.1 | - | 9 | 1694 | ||
| AK041743.1 | - | 9 | 1239 | |||
| AK048626.1 | - | 9 | 1718 | |||
| AK083158.1 | - | 9 | 1326 | |||
| AK210600.1 | - | 1 | 88 | |||
| BC066850.1 | - | 9 | 1404 | 30432720 | ||

Smad3
Genomic Map