siRNA Details
| Gene Symbol: Nasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081475.1 | NP_001074944.1 | 5 | 449 | ||
| NM_001284229.1 | NP_001271158.1 | 4 | 368 | |||
| NM_016777.3 | NP_058057.3 | 6 | 443 | |||
| GenBank mRNA | AF034610.2 | - | 6 | 433 | ||
| AF095722.1 | - | 4 | 349 | |||
| AK083333.1 | - | 5 | 450 | |||
| AK132690.1 | - | 6 | 422 | |||
| AK161367.1 | - | 6 | 422 | |||
| BC004693.1 | - | 5 | 416 | 3498591 | ||

Nasp
Genomic Map