siRNA Details
| Gene Symbol: Nptx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016789.3 | NP_058069.1 | 5 | 1337 | ||
| GenBank mRNA | AF049124.1 | - | 5 | 1227 | ||
| AK134806.1 | - | 5 | 1338 | |||
| BC026054.1 | - | 5 | 1332 | 4038070 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nptx2| Gene Symbol: Nptx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016789.3 | NP_058069.1 | 5 | 1337 | ||
| GenBank mRNA | AF049124.1 | - | 5 | 1227 | ||
| AK134806.1 | - | 5 | 1338 | |||
| BC026054.1 | - | 5 | 1332 | 4038070 | ||