siRNA Details
| Gene Symbol: Incenp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016692.3 | NP_057901.2 | 12 | 1906 | ||
| GenBank mRNA | AB100432.1 | - | 11 | 1700 | ||
| AB100433.1 | - | 12 | 1712 | |||
| AF117610.1 | - | 12 | 1842 | |||
| AK012497.1 | - | 2 | 169 | |||
| AK034764.1 | - | 12 | 1835 | |||
| AK081841.1 | - | 12 | 1840 | |||
| AK088627.1 | - | 12 | 1863 | |||
| BC052414.1 | - | 12 | 1839 | 6401188 | ||

Incenp
Genomic Map