siRNA Details
| Gene Symbol: Mapk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016700.4 | NP_057909.1 | 8 | 969 | ||
| GenBank mRNA | AB005663.1 | - | 7 | 802 | ||
| AB206575.1 | - | 7 | 744 | |||
| AB219977.1 | - | 7 | 778 | |||
| AK043102.1 | - | 8 | 969 | |||
| AK139498.1 | - | 8 | 957 | |||
| BC053027.1 | - | 8 | 939 | 5694771 | ||

Mapk8
Genomic Map