siRNA Details
| Gene Symbol: Agxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276710.1 | NP_001263639.1 | 5, 6 | 723 | ||
| NM_016702.3 | NP_057911.2 | 5, 6 | 723 | |||
| GenBank mRNA | AF027730.2 | - | 5, 6 | 718 | ||
| AK149432.1 | - | 5, 6 | 646 | |||
| BC025799.1 | - | 5, 6 | 687 | 5099627 | ||

Agxt
Genomic Map