siRNA Details
| Gene Symbol: Preb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294302.1 | NP_001281231.1 | 5 | 902 | ||
| NM_016703.3 | NP_057912.2 | 5 | 902 | |||
| GenBank mRNA | AF150808.1 | - | 5 | 805 | ||
| AF193017.1 | - | 5 | 814 | |||
| AK013330.1 | - | 5 | 856 | |||
| AK029838.1 | - | 5 | 838 | |||
| AK036866.1 | - | 5 | 840 | |||
| AK151288.1 | - | 5 | 884 | |||
| AK188795.1 | - | 3 | 215 | |||
| AK213634.1 | - | 3 | 217 | |||
| BC017527.1 | - | 5 | 805 | 4921119 | ||

Preb
Genomic Map