siRNA Details
| Gene Symbol: Auh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016709.2 | NP_057918.2 | 10 | 1229 | ||
| GenBank mRNA | AK003219.1 | - | 10 | 1209 | ||
| BB207692.1 | - | 1 | 216 | |||
| BC026525.1 | - | 10 | 1167 | |||
| BC049597.1 | - | 10 | 1213 | 6593786 | ||

Auh
Genomic Map