siRNA Details
| Gene Symbol: Sult1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016771.3 | NP_058051.3 | 8 | 1294 | ||
| GenBank mRNA | AF026073.1 | - | 8 | 1172 | ||
| AK017162.1 | - | 2 | 488 | |||
| AK133530.1 | - | 8 | 1213 | |||
| BC010752.1 | - | 8 | 1268 | |||
| BC066190.1 | - | 8 | 1247 | 30472198 | ||

Sult1d1
Genomic Map