siRNA Details
| Gene Symbol: Nasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081475.1 | NP_001074944.1 | 13 | 1357 | ||
| NM_001284229.1 | NP_001271158.1 | 12 | 1276 | |||
| NM_016777.3 | NP_058057.3 | 15 | 2326 | |||
| GenBank mRNA | AF034610.2 | - | 15 | 2316 | ||
| AF095722.1 | - | 12 | 1257 | |||
| AK083333.1 | - | 13 | 1358 | |||
| BC004693.1 | - | 13 | 1324 | 3498591 | ||

Nasp
Genomic Map