siRNA Details
| Gene Symbol: GJC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080383.1 | NP_001073852.1 | 3 | 396 | ||
| NM_005497.3 | NP_005488.2 | 3 | 400 | |||
| XM_005256920.1 | XP_005256977.1 | 3 | 394 | |||
| XM_005256921.1 | XP_005256978.1 | 2 | 222 | |||
| GenBank mRNA | AK124339.1 | - | 2 | 634 | ||
| BC096213.3 | - | 1 | 126 | 40004800 | ||
| BC096214.3 | - | 1 | 126 | 40004801 | ||
| BC096215.3 | - | 1 | 126 | 40004803 | ||
| BC096216.3 | - | 1 | 126 | 40004809 | ||
| CN299428.1 | - | 3 | 414 | |||
| DA845933.1 | - | 3 | 396 | |||

GJC1
Genomic Map