siRNA Details
| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016784.3 | NP_058064.2 | 3, 4 | 375 | ||
| GenBank mRNA | AF044334.1 | - | 2, 3 | 267 | ||
| AK075993.1 | - | 3, 4 | 363 | |||
| AK076007.1 | - | 3, 4 | 353 | |||
| AK077197.1 | - | 3, 4 | 376 | |||
| AK080068.1 | - | 3, 4 | 382 | |||
| BC006750.1 | - | 3, 4 | 345 | 3709134 | ||
| BC052895.1 | - | 3, 4 | 333 | |||

Plrg1
Genomic Map