siRNA Details
| Gene Symbol: Tpmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016785.2 | NP_058065.2 | 6 | 620 | ||
| GenBank mRNA | AF037043.1 | - | 6 | 529 | ||
| AF037044.1 | - | 5 | 480 | |||
| AF046887.1 | - | 6 | 606 | |||
| AK002335.1 | - | 6 | 527 | |||
| BC021598.1 | - | 5 | 507 | 5098857 | ||

Tpmt
Genomic Map