siRNA Details
| Gene Symbol: Ube2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016786.3 | NP_058066.2 | 7 | 2542 | ||
| GenBank mRNA | AK031055.1 | - | 5 | 2242 | ||
| AK048194.1 | - | 7 | 2532 | |||
| AK149463.1 | - | 1 | 468 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2k| Gene Symbol: Ube2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016786.3 | NP_058066.2 | 7 | 2542 | ||
| GenBank mRNA | AK031055.1 | - | 5 | 2242 | ||
| AK048194.1 | - | 7 | 2532 | |||
| AK149463.1 | - | 1 | 468 | |||