siRNA Details
| Gene Symbol: Rps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016844.2 | NP_058540.1 | 2, 3 | 121 | ||
| GenBank mRNA | AK002919.1 | - | 2, 3 | 119 | ||
| AV497904.1 | - | 2, 3 | 100 | |||
| BC010987.1 | - | 2, 3 | 116 | 4218716 | ||
| BC090982.1 | - | 2, 3 | 118 | 30316901 | ||
| BC125644.1 | - | 2, 3 | 185 | 40130159 | ||
| BC125646.1 | - | 2, 3 | 185 | 40130161 | ||
| BY502260.1 | - | 2, 3 | 124 | |||
| U11248.1 | - | 2, 3 | 102 | |||

Rps28
Genomic Map