siRNA Details
| Gene Symbol: Rab25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016899.4 | NP_058595.2 | 3 | 536 | ||
| GenBank mRNA | AB232620.1 | - | 3 | 332 | ||
| AF119675.1 | - | 3 | 472 | |||
| AK003260.1 | - | 3 | 537 | |||
| BC006624.1 | - | 3 | 522 | 3968810 | ||

Rab25
Genomic Map