siRNA Details
| Gene Symbol: Cdkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276315.1 | NP_001263244.1 | 4 | 1017 | ||
| NM_016912.2 | NP_058608.1 | 4 | 751 | |||
| NM_177270.4 | NP_796244.2 | 4 | 751 | |||
| GenBank mRNA | AB029065.1 | - | 4 | 727 | ||
| AB029066.1 | - | 4 | 715 | |||
| AB029067.1 | - | 4 | 1005 | |||
| AK030598.1 | - | 4 | 1006 | |||
| AK139820.1 | - | 4 | 959 | |||
| AK144574.1 | - | 4 | 752 | |||
| AK145688.1 | - | 4 | 900 | |||
| BC117894.1 | - | 4 | 660 | 40096852 | ||
| BC117895.1 | - | 4 | 660 | 40096853 | ||

Cdkl2
Genomic Map