siRNA Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016843.3 | NP_058539.2 | 6, 7 | 815 | ||
| GenBank mRNA | AK002504.1 | - | 6, 7 | 924 | ||
| AK003530.1 | - | 6, 7 | 742 | |||
| AK047170.1 | - | 6, 7 | 865 | |||
| AK049706.1 | - | 6, 7 | 859 | |||
| AK077738.1 | - | 6, 7 | 817 | |||
| AK078459.1 | - | 6, 7 | 895 | |||
| AK153271.1 | - | 6, 7 | 817 | |||
| AK161950.1 | - | 832 | ||||
| AK165941.1 | - | 6, 7 | 840 | |||
| AK166899.1 | - | 6, 7 | 900 | |||
| AK170507.1 | - | 6, 7 | 832 | |||
| AK170751.1 | - | 6, 7 | 841 | |||
| AK195052.1 | - | 1, 2 | 49 | |||
| AK219896.1 | - | 1, 2 | 49 | |||
| BC016410.1 | - | 6, 7 | 825 | 3968016 | ||
| BC046802.1 | - | 6, 7 | 951 | 6404607 | ||
| X61506.1 | - | 6, 7 | 929 | |||

Atxn10
Genomic Map