siRNA Details
| Gene Symbol: Wbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016852.2 | NP_058548.1 | 3 | 281 | ||
| GenBank mRNA | AK004824.1 | - | 3 | 283 | ||
| AK043037.1 | - | 3 | 255 | |||
| AK146572.1 | - | 3 | 283 | |||
| AK150018.1 | - | 3 | 256 | |||
| AK151368.1 | - | 3 | 273 | |||
| AK162810.1 | - | 3 | 256 | |||
| AK170317.1 | - | 3 | 282 | |||
| AK170474.1 | - | 3 | 284 | |||
| BC031908.1 | - | 3 | 244 | 5365118 | ||
| BC055058.1 | - | 3 | 301 | 6529458 | ||
| U40826.1 | - | 3 | 281 | |||

Wbp2
Genomic Map