siRNA Details
| Gene Symbol: Bysl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016859.3 | NP_058555.3 | 4 | 932 | ||
| GenBank mRNA | AF007802.1 | - | 2 | 161 | ||
| AK019239.1 | - | 4 | 667 | |||
| AK032993.1 | - | 4 | 664 | |||
| AK049195.1 | - | 4 | 925 | |||
| AK051704.1 | - | 4 | 922 | |||
| AK083033.1 | - | 4 | 919 | |||
| AK143027.1 | - | 4 | 933 | |||
| AK168301.1 | - | 4 | 670 | |||
| BC017530.1 | - | 4 | 659 | 4925307 | ||

Bysl
Genomic Map