siRNA Details
| Gene Symbol: Ramp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016894.3 | NP_058590.1 | 3 | 930 | ||
| GenBank mRNA | AB019046.1 | - | 3 | 741 | ||
| AF146522.1 | - | 3 | 722 | |||
| AF209904.1 | - | 3 | 701 | |||
| AJ250489.1 | - | 3 | 722 | |||
| AK004539.1 | - | 1 | 348 | |||
| AK157896.1 | - | 3 | 720 | |||
| AK165238.1 | - | 3 | 706 | |||
| BC012644.1 | - | 3 | 687 | 4207524 | ||

Ramp1
Genomic Map