siRNA Details
| Gene Symbol: Cd164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016898.2 | NP_058594.1 | 6 | 574 | ||
| GenBank mRNA | AB014464.1 | - | 6 | 539 | ||
| AB028895.1 | - | 6 | 574 | |||
| AF299345.1 | - | 6 | 539 | |||
| AK002792.1 | - | 1 | 20 | |||
| AK006592.1 | - | 6 | 433 | |||
| AK145131.1 | - | 6 | 633 | |||
| AK169275.1 | - | 6 | 614 | |||
| BC005414.1 | - | 6 | 562 | 3600535 | ||

Cd164
Genomic Map