siRNA Details
| Gene Symbol: Rwdd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016924.2 | NP_058620.2 | 4 | 724 | ||
| GenBank mRNA | AF177771.1 | - | 4 | 680 | ||
| AK002746.1 | - | 4 | 690 | |||
| AK084966.1 | - | 4 | 699 | |||
| BC002143.1 | - | 4 | 705 | 3484719 | ||

Rwdd2b
Genomic Map