siRNA Details
| Gene Symbol: Dgka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016811.2 | NP_058091.2 | 18 | 1761 | ||
| GenBank mRNA | AF085219.1 | - | 17 | 1447 | ||
| AK008063.1 | - | 6 | 425 | |||
| AK008314.1 | - | 7 | 432 | |||
| BC006713.1 | - | 18 | 1761 | 3584592 | ||
| JN832677.1 | - | 18 | 1704 | |||
| JN832678.1 | - | 18 | 1809 | |||
| JN832679.1 | - | 18 | 1780 | |||

Dgka
Genomic Map