siRNA Details
| Gene Symbol: Pmm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016881.2 | NP_058577.1 | 7 | 620 | ||
| GenBank mRNA | AF043514.1 | - | 7 | 620 | ||
| AK002858.1 | - | 7 | 562 | |||
| AK003334.1 | - | 7 | 578 | |||
| AK005074.1 | - | 7 | 602 | |||
| AK134850.1 | - | 572 | ||||
| AK145402.1 | - | 7 | 572 | |||
| AK149116.1 | - | 7 | 582 | |||
| BC037101.1 | - | 7 | 597 | |||
| BC046325.1 | - | 7 | 568 | 6395635 | ||
| BC094448.1 | - | 7 | 586 | 4238664 | ||

Pmm2
Genomic Map