siRNA Details
| Gene Symbol: KCNJ10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002241.4 | NP_002232.2 | 2 | 477 | ||
| GenBank mRNA | AK312383.1 | - | 2 | 384 | ||
| BC034036.1 | - | 2 | 379 | |||
| BC131627.1 | - | 2 | 373 | 40118533 | ||
| DA958759.1 | - | 2 | 477 | |||
| U52155.1 | - | 2 | 463 | |||

KCNJ10
Genomic Map